Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
snt-1
Entrez Gene ID
174120
Gene Name
protein-coding
Gene Aliases
CELE_F31E8.2
Chromosome Location
Chr.II: 6810963 - 6816463 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026958.5 NP_001022129.1 5 - 6 986 89
GenBank mRNA L15302.1 - 5 - 6 1008 89

View Full Product Details