Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
npp-3
Entrez Gene ID
174823
Gene Name
Nuclear Pore complex Protein
Gene Aliases
CELE_K12D12.2
Chromosome Location
Chr.II: 11865431 - 11874788 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064133.5 NP_496534.1 2 - 3 585 110

View Full Product Details