Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- dct-18
- Entrez Gene ID
-
174934
- Gene Name
- DAF-16/FOXO Controlled, germline Tumor affecting
- Gene Aliases
-
CELE_F58G1.4
- Chromosome Location
-
Chr.II: 12930525 - 12933524 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_064354.7 |
NP_496755.1 |
2 - 3 |
398 |
63 |
View Full Product Details