Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ccdc-47
Entrez Gene ID
175505
Gene Name
CCDC (human Coiled Coil Domain Containing) homolog
Gene Aliases
CELE_ZK1058.4
Chromosome Location
Chr.III: 3916967 - 3918814 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065387.4 NP_497788.1 2 - 3 473 57

View Full Product Details