Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hum-5
Entrez Gene ID
175521
Gene Name
Heavy chain, Unconventional Myosin
Gene Aliases
CELE_T02C12.1
Chromosome Location
Chr.III: 4029115 - 4036834 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065408.8 NP_497809.1 10 - 11 1839 90
GenBank mRNA X75564.1 - - 1850 90

View Full Product Details