Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-10
Entrez Gene ID
191836
Gene Name
Serpentine receptor class gamma-10
Gene Aliases
CELE_T04A8.1
Chromosome Location
Chr.III: 4677766 - 4679663 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065553.1 NP_497954.1 4 - 5 775 66

View Full Product Details