Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dct-6
Entrez Gene ID
175846
Gene Name
protein-coding
Gene Aliases
CELE_F25B5.1
Chromosome Location
Chr.III: 5966413 - 5971519 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038268.4 NP_001033357.2 4 - 5 522 114
NM_001038269.5 NP_001033358.2 4 - 5 522 114

View Full Product Details