Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cgh-1
Entrez Gene ID
176061
Gene Name
ATP-dependent RNA helicase cgh-1
Gene Aliases
CELE_C07H6.5
Chromosome Location
Chr.III: 7495320 - 7497834 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066245.7 NP_498646.1 3 - 4 1065 80

View Full Product Details