Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dnj-10
Entrez Gene ID
176211
Gene Name
protein-coding
Gene Aliases
CELE_F22B7.5
Chromosome Location
Chr.III: 8631165 - 8633565 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066501.4 NP_498902.1 7 - 8 1261 83

View Full Product Details