Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cut-6
Entrez Gene ID
176520
Gene Name
CUTiclin
Gene Aliases
CELE_M142.2
Chromosome Location
Chr.III: 10911478 - 10916665 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066999.7 NP_499400.2 3 - 4 343 80

View Full Product Details