Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cut-6
Entrez Gene ID
176520
Gene Name
CUTiclin
Gene Aliases
CELE_M142.2
Chromosome Location
Chr.III: 10911478 - 10916665 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066999.7 NP_499400.2 5 - 6 902 156

View Full Product Details