Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tbx-33
Entrez Gene ID
190493
Gene Name
Putative T-box protein 33
Gene Aliases
CELE_Y66A7A.8
Chromosome Location
Chr.III: 11633214 - 11636840 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067105.5 NP_499506.2 1 - 2 192 67

View Full Product Details