Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ndc-80
Entrez Gene ID
177873
Gene Name
Kinetochore protein ndc-80
Gene Aliases
CELE_W01B6.9
Chromosome Location
Chr.IV: 10086137 - 10088437 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069429.5 NP_501830.1 1 - 2 118 100

View Full Product Details