Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
elt-6
Entrez Gene ID
176993
Gene Name
GATA-type domain-containing protein
Gene Aliases
CELE_F52C12.5
Chromosome Location
Chr.IV: 1917977 - 1921646 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067743.7 NP_500144.2 6 - 7 968 72
GenBank mRNA AF353303.1 - 6 - 7 968 72

View Full Product Details