Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
clec-70
Entrez Gene ID
189944
Gene Name
C-type lectin domain-containing protein
Gene Aliases
CELE_Y46C8AL.3
Chromosome Location
Chr.IV: 3932708 - 3936307 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068038.4 NP_500439.2 1 - 2 238 83

View Full Product Details