Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srm-1
Entrez Gene ID
191955
Gene Name
Serpentine Receptor, class M
Gene Aliases
CELE_T28H11.2
Chromosome Location
Chr.IV: 5012541 - 5014842 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068301.3 NP_500702.1 5 - 6 917 101

View Full Product Details