Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tag-89
Entrez Gene ID
178062
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_H02I12.3
Chromosome Location
Chr.IV: 11394543 - 11396818 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069746.2 NP_502147.1 2 - 3 312 135

View Full Product Details