Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srz-15
Entrez Gene ID
183127
Gene Name
Serpentine Receptor, class Z
Gene Aliases
CELE_C32H11.2
Chromosome Location
Chr.IV: 12915779 - 12917293 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070063.2 NP_502464.1 2 - 3 542 185

View Full Product Details