Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ccg-1
Entrez Gene ID
178426
Gene Name
Conserved Cysteine/Glycine domain protein
Gene Aliases
CELE_Y73F8A.6
Chromosome Location
Chr.IV: 15270160 - 15275835 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070441.7 NP_502842.1 2 - 3 751 66

View Full Product Details