Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lgc-26
Entrez Gene ID
178554
Gene Name
Neur_chan_LBD domain-containing protein
Gene Aliases
CELE_Y58G8A.1
Chromosome Location
Chr.V: 268238 - 271419 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070766.4 NP_503167.1 5 - 6 736 107

View Full Product Details