Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
unc-34
Entrez Gene ID
190098
Gene Name
protein-coding
Gene Aliases
CELE_Y50D4C.1
Chromosome Location
Chr.V: 982706 - 996914 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313345.3 NP_001300274.1 4 - 4 1115 133
NM_001313346.3 NP_001300275.1 3 - 4 722 133
NM_001313347.3 NP_001300276.1 2 - 3 368 133
NM_001313348.3 NP_001300277.1 1 - 2 230 133
NM_070959.8 NP_503360.4 4 - 5 1145 133
GenBank mRNA AF535160.1 - 5 - 6 1340 133

View Full Product Details