Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srj-50
Entrez Gene ID
191951
Gene Name
Serpentine Receptor, class J
Gene Aliases
CELE_T03D3.2
Chromosome Location
Chr.V: 2832966 - 2834921 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071366.1 NP_503767.1 3 - 4 654 80

View Full Product Details