Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-35A5
Entrez Gene ID
178802
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_K07C6.5
Chromosome Location
Chr.V: 3936326 - 3938597 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071691.7 NP_504092.1 4 - 5 828 108

View Full Product Details