Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gpa-10
Entrez Gene ID
179075
Gene Name
Guanine nucleotide-binding protein alpha-10 subunit
Gene Aliases
CELE_C55H1.2
Chromosome Location
Chr.V: 6852521 - 6854811 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072344.3 NP_504745.1 4 - 5 473 80
GenBank mRNA AY008132.1 - 4 - 5 465 80

View Full Product Details