Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ech-3
Entrez Gene ID
179180
Gene Name
protein-coding
Gene Aliases
CELE_F43H9.1
Chromosome Location
Chr.V: 8022217 - 8023801 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322576.3 NP_001309646.1 1 - 2 130 123
NM_072665.6 NP_505066.1 2 - 3 435 123

View Full Product Details