Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srp-8
Entrez Gene ID
191960
Gene Name
SERPIN domain-containing protein
Gene Aliases
CELE_F20D6.3
Chromosome Location
Chr.V: 8183684 - 8185879 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322578.3 NP_001309497.1 4 - 5 848 117

View Full Product Details