Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srsx-31
Entrez Gene ID
3565162
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_F07B10.6
Chromosome Location
Chr.V: 11253872 - 11256693 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028596.2 NP_001023767.1 4 - 5 836 94

View Full Product Details