Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ape-1
Entrez Gene ID
179601
Gene Name
protein-coding
Gene Aliases
CELE_F46F3.4
Chromosome Location
Chr.V: 11675938 - 11685274 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269424.2 NP_001256353.1 7 - 8 2276 70
NM_001269425.4 NP_001256354.1 5 - 6 2060 70
NM_001269426.3 NP_001256355.1 2 - 3 1244 70
NM_001269427.3 NP_001256356.1 1 - 2 968 70

View Full Product Details