Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srj-4
Entrez Gene ID
182952
Gene Name
Serpentine Receptor, class J
Gene Aliases
CELE_C27A7.7
Chromosome Location
Chr.V: 12163206 - 12164535 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073650.1 NP_506051.1 3 - 4 820 84

View Full Product Details