Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cdr-7
Entrez Gene ID
24104698
Gene Name
protein-coding
Gene Aliases
CELE_K01D12.13
Chromosome Location
Chr.V: 12410879 - 12412113 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307735.3 NP_001294664.1 4 - 5 514 144
NM_001307795.3 NP_001294724.1 4 - 5 438 144
GenBank mRNA AY728061.1 - 4 - 5 467 144

View Full Product Details