Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ril-2
Entrez Gene ID
179725
Gene Name
protein-coding
Gene Aliases
CELE_C14C10.3
Chromosome Location
Chr.V: 12596169 - 12599157 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028487.7 NP_001023658.1 4 - 5 1106 112
NM_001028488.6 NP_001023659.1 4 - 5 1108 112
NM_001269507.4 NP_001256436.1 4 - 5 1112 112

View Full Product Details