Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wrt-8
Entrez Gene ID
180035
Gene Name
Warthog protein 8 N-product
Gene Aliases
CELE_C29F3.2
Chromosome Location
Chr.V: 15342054 - 15344581 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074404.6 NP_506805.1 3 - 4 469 63

View Full Product Details