Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- pptr-1
- Entrez Gene ID
-
180100
- Gene Name
- Serine/threonine-protein phosphatase 2A regulatory subunit pptr-1
- Gene Aliases
-
CELE_W08G11.4
- Chromosome Location
-
Chr.V: 16357521 - 16367574 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_074732.6 |
NP_507133.4 |
4 - 5 |
889 |
78 |
View Full Product Details