Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ife-4
Entrez Gene ID
180464
Gene Name
Eukaryotic translation initiation factor 4E-4
Gene Aliases
CELE_C05D9.5
Chromosome Location
Chr.X: 1104212 - 1106251 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075809.5 NP_508210.1 2 - 3 242 93
GenBank mRNA AF214651.1 - 2 - 3 242 93

View Full Product Details