Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
calu-1
Entrez Gene ID
180769
Gene Name
protein-coding
Gene Aliases
CELE_M03F4.7
Chromosome Location
Chr.X: 4969529 - 4972540 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383568.1 5 - 6 1186 66
NM_001383569.1 2 - 3 815 66
GenBank mRNA AF228528.1 - 3 - 4 776 66

View Full Product Details