Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ifta-1
Entrez Gene ID
183817
Gene Name
WD_REPEATS_REGION domain-containing protein
Gene Aliases
CELE_C54G7.4
Chromosome Location
Chr.X: 5545587 - 5552756 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076539.6 NP_508940.4 13 - 14 1743 114

View Full Product Details