Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
irk-1
Entrez Gene ID
180944
Gene Name
Inward rectifier potassium channel irk-1
Gene Aliases
CELE_R03E9.4
Chromosome Location
Chr.X: 6765970 - 6778114 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076737.5 NP_509138.2 3 - 4 470 84

View Full Product Details