Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rhgf-1
Entrez Gene ID
184419
Gene Name
RHo Guanine nucleotide exchange Factor
Gene Aliases
CELE_F13E6.6
Chromosome Location
Chr.X: 10697834 - 10711869 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077390.10 NP_509791.4 22 - 23 3252 62

View Full Product Details