Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oct-2
Entrez Gene ID
181325
Gene Name
protein-coding
Gene Aliases
CELE_ZK455.8
Chromosome Location
Chr.X: 11346455 - 11350247 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029821.6 NP_001024992.2 2 - 3 425 123
NM_077498.9 NP_509899.3 7 - 8 1276 123
GenBank mRNA AF199013.1 - 2 - 3 425 123

View Full Product Details