Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-40
Entrez Gene ID
184608
Gene Name
Serpentine receptor class delta-40
Gene Aliases
CELE_F17A2.12
Chromosome Location
Chr.X: 12323739 - 12325668 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077661.2 NP_510062.1 5 - 6 788 121

View Full Product Details