Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gfi-3
Entrez Gene ID
181544
Gene Name
Protein gfi-3
Gene Aliases
CELE_M163.4
Chromosome Location
Chr.X: 14480572 - 14484702 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078008.7 NP_510409.1 1 - 2 114 86

View Full Product Details