Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-3
Entrez Gene ID
181551
Gene Name
Nuclear hormone receptor family member nhr-3
Gene Aliases
CELE_H01A20.1
Chromosome Location
Chr.X: 14558504 - 14561432 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078022.7 NP_510423.1 2 - 3 305 129
GenBank mRNA AF083222.1 - 2 - 3 316 129
AF273769.1 - 2 - 3 312 129

View Full Product Details