Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srw-41
Entrez Gene ID
3564767
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_F14F8.10
Chromosome Location
Chr.V: 16697478 - 16699773 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028627.1 NP_001023798.1 1 - 2 111 155

View Full Product Details