Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxb-90
Entrez Gene ID
4363043
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_F12E12.10
Chromosome Location
Chr.II: 3746172 - 3748819 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047296.4 NP_001040761.1 1 - 2 104 118

View Full Product Details