Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDUFA4
Entrez Gene ID
100856334
Gene Name
NDUFA4 mitochondrial complex associated
Gene Aliases
-
Chromosome Location
Chr.14: 25887204 - 25893746 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003639479.4 XP_003639527.1 3 - 4 334 77

View Full Product Details