Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZNRD2
Entrez Gene ID
476024
Gene Name
zinc ribbon domain containing 2
Gene Aliases
SSSCA1
Chromosome Location
Chr.18: 51634946 - 51636330 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014121011.2 XP_013976486.1 2 - 3 453 53
XM_533233.6 XP_533233.2 3 - 4 389 53

View Full Product Details