Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLK15
Entrez Gene ID
480822
Gene Name
kallikrein related peptidase 15
Gene Aliases
-
Chromosome Location
Chr.1: 106005601 - 106007583 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022423219.1 XP_022278927.1 1 - 2 307 57

View Full Product Details