Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNRPD1
Entrez Gene ID
480178
Gene Name
small nuclear ribonucleoprotein D1 polypeptide
Gene Aliases
-
Chromosome Location
Chr.7: 66407313 - 66421193 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287157.1 NP_001274086.1 1 - 2 131 64
GenBank mRNA CO657835.1 - 3 - 4 460 64
CO672980.1 - 3 - 4 465 64
DT536786.1 - 1 - 2 117 64

View Full Product Details