Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPEM1
Entrez Gene ID
607805
Gene Name
spermatid maturation 1
Gene Aliases
-
Chromosome Location
Chr.5: 32354044 - 32357684 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_844610.5 XP_849703.1 2 - 3 302 73

View Full Product Details