Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INHBE
Entrez Gene ID
100856447
Gene Name
inhibin subunit beta E
Gene Aliases
-
Chromosome Location
Chr.10: 1558876 - 1563370 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005625539.3 XP_005625596.1 - 232 58

View Full Product Details